siRNA Details
| Gene Symbol: HOXB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024015.4 | NP_076920.1 | 2 | 1181 | ||
| GenBank mRNA | AL137449.1 | - | 2 | 1105 | ||
| BC049204.1 | - | 2 | 1181 | 5533346 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXB4| Gene Symbol: HOXB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024015.4 | NP_076920.1 | 2 | 1181 | ||
| GenBank mRNA | AL137449.1 | - | 2 | 1105 | ||
| BC049204.1 | - | 2 | 1181 | 5533346 | ||