siRNA Details
| Gene Symbol: MMP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002428.3 | NP_002419.1 | 4 | 1372 | ||
| GenBank mRNA | BC006412.1 | - | 4 | 492 | ||
| BC036495.1 | - | 4 | 1378 | 5263145 | ||
| BC055428.1 | - | 4 | 1373 | 4811572 | ||
| BT006715.1 | - | 2 | 272 | |||
| D85510.1 | - | 1 | 102 | |||
| D86331.1 | - | 3 | 310 | |||
| Z48482.1 | - | 4 | 635 | |||

MMP15
Genomic Map