siRNA Details
| Gene Symbol: PGLYRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052890.3 | NP_443122.3 | 2 | 1197 | ||
| XM_006722633.3 | XP_006722696.1 | 2 | 1194 | |||
| GenBank mRNA | AB073610.1 | - | 2 | 1146 | ||
| AF384856.1 | - | 2 | 1090 | |||
| AK055882.1 | - | 2 | 1194 | |||
| AK289415.1 | - | 2 | 1130 | |||
| AK292292.1 | - | 2 | 1545 | |||
| AY358156.1 | - | 2 | 1121 | |||
| BC136551.1 | - | 2 | 1086 | 9020541 | ||
| BC136552.1 | - | 2 | 1086 | 9020542 | ||
| BC144238.1 | - | 2 | 1086 | 9052758 | ||

PGLYRP2
Genomic Map