siRNA Details
| Gene Symbol: RAB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317915.1 | NP_001304844.1 | 5 | 846 | ||
| NM_138453.3 | NP_612462.1 | 5 | 808 | |||
| GenBank mRNA | AY026936.1 | - | 5 | 766 | ||
| BC013033.2 | - | 5 | 740 | 3534915 | ||

RAB3C
Genomic Map