siRNA Details
| Gene Symbol: ESX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153448.3 | NP_703149.1 | 4 | 673 | ||
| GenBank mRNA | AK097704.1 | - | 4 | 688 | ||
| AY114148.1 | - | 4 | 647 | |||
| BC042633.1 | - | 4 | 634 | 5170565 | ||
| BC053599.1 | - | 4 | 662 | 5742902 | ||

ESX1
Genomic Map