siRNA Details
| Gene Symbol: PASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173493.2 | NP_775764.2 | 9, 10 | 1057 | ||
| XM_011531102.1 | XP_011529404.1 | 9, 10 | 970 | |||
| GenBank mRNA | AK097552.1 | - | 9, 10 | 970 | ||
| AY270020.1 | - | 9, 10 | 863 | |||
| BC040301.1 | - | 9, 10 | 868 | 4826573 | ||

PASD1
Genomic Map