siRNA Details
| Gene Symbol: MORF4L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001265603.1 | NP_001252532.1 | 10 | 1677 | ||
| NM_001265604.1 | NP_001252533.1 | 12 | 1745 | |||
| NM_001265605.1 | NP_001252534.1 | 12 | 1645 | |||
| NM_006791.3 | NP_006782.1 | 12 | 1792 | |||
| NM_206839.2 | NP_996670.1 | 13 | 1909 | |||
| GenBank mRNA | AF070664.1 | - | 12 | 1599 | ||
| AF100615.1 | - | 12 | 1686 | |||
| AF109188.1 | - | 12 | 1582 | |||
| AF131847.1 | - | 8 | 1315 | |||
| AF161546.1 | - | 12 | 1581 | |||
| AF167173.1 | - | 12 | 1594 | |||
| AF218011.1 | - | 12 | 1712 | |||
| AK095823.1 | - | 13 | 2194 | |||
| AK098485.1 | - | 12 | 1716 | |||
| AL137697.1 | - | 12 | 1587 | |||
| AY148481.1 | - | 13 | 1803 | |||
| BC002936.2 | - | 12 | 1674 | 3955120 | ||
| BC022845.2 | - | 12 | 1674 | 2988803 | ||
| BC067826.1 | - | 12 | 1569 | 4824022 | ||
| BC141840.1 | - | 12 | 1600 | 8860303 | ||

MORF4L1
Genomic Map