siRNA Details
| Gene Symbol: MEFV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000243.2 | NP_000234.1 | 10 | 2231 | ||
| NM_001198536.1 | NP_001185465.1 | 9 | 1773 | |||
| XM_017023236.1 | XP_016878725.1 | 10 | 2369 | |||
| GenBank mRNA | AF018080.1 | - | 10 | 2232 | ||
| BC101511.1 | - | 10 | 2216 | 8069017 | ||
| BC101537.1 | - | 10 | 2216 | 8069043 | ||
| FJ785717.1 | - | 7 | 1112 | |||
| FJ785718.1 | - | 6 | 996 | |||
| FJ785719.1 | - | 5 | 963 | |||
| FJ785720.1 | - | 6 | 881 | |||
| FJ785721.1 | - | 6 | 1596 | |||
| FJ785722.1 | - | 9 | 1735 | |||
| Y14441.1 | - | 10 | 2220 | |||

MEFV
Genomic Map