siRNA Details
Gene Symbol: RFXAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000538.3 | NP_000529.1 | 1 | 437 | ||
GenBank mRNA | BC026088.1 | - | 1 | 416 | 4827378 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RFXAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000538.3 | NP_000529.1 | 1 | 437 | ||
GenBank mRNA | BC026088.1 | - | 1 | 416 | 4827378 |