siRNA Details
| Gene Symbol: CDC5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253.3 | NP_001244.1 | 2, 3 | 460 | ||
| GenBank mRNA | AB007892.2 | - | 2, 3 | 232 | ||
| AK128737.1 | - | 2, 3 | 268 | |||
| AK299738.1 | - | 2, 3 | 268 | |||
| BC001568.1 | - | 2, 3 | 249 | 3452352 | ||
| BG497404.1 | - | 411 | ||||
| BM829346.1 | - | 2, 3 | 460 | |||
| D85423.1 | - | 1, 2 | 129 | |||
| DC422730.1 | - | 2, 3 | 459 | |||
| U86753.1 | - | 2, 3 | 150 | |||

CDC5L
Genomic Map