siRNA Details
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001537.3 | NP_001528.1 | 4 | 607 | ||
| GenBank mRNA | AF068754.1 | - | 4 | 487 | ||
| AK172732.1 | - | 4 | 472 | |||
| AY320409.1 | - | 4 | 492 | |||
| BC007515.2 | - | 4 | 479 | 3010737 | ||
| BX366956.2 | - | 4 | 539 | |||
| BX537440.1 | - | 4 | 494 | |||
| BX648972.1 | - | 4 | 491 | |||

HSBP1
Genomic Map