siRNA Details
Gene Symbol: MN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002430.2 | NP_002421.3 | 2 | 6463 | ||
GenBank mRNA | X82209.2 | - | 2 | 6462 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002430.2 | NP_002421.3 | 2 | 6463 | ||
GenBank mRNA | X82209.2 | - | 2 | 6462 |