siRNA Details
| Gene Symbol: MYBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278610.1 | NP_001265539.1 | 8, 9 | 1692 | ||
| NM_002466.3 | NP_002457.1 | 9, 10 | 1764 | |||
| GenBank mRNA | AK223482.1 | - | 9, 10 | 1499 | ||
| AK303249.1 | - | 8, 9 | 1597 | |||
| AK314791.1 | - | 9, 10 | 1669 | |||
| BC007585.1 | - | 9, 10 | 1689 | 3162656 | ||
| BC053555.1 | - | 9, 10 | 1658 | 6140320 | ||
| X13293.1 | - | 9, 10 | 1626 | |||

MYBL2
Genomic Map