siRNA Details
| Gene Symbol: DTYMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165031.1 | NP_001158503.1 | 3 | 445 | ||
| NM_001320905.1 | NP_001307834.1 | 4 | 562 | |||
| NM_012145.3 | NP_036277.2 | 3 | 445 | |||
| NR_135492.1 | 3 | 407 | ||||
| GenBank mRNA | AK223434.1 | - | 3 | 292 | ||
| BC001827.2 | - | 3 | 285 | 3533327 | ||
| BC147006.1 | - | 3 | 384 | 9056934 | ||
| BC147010.1 | - | 3 | 384 | 9056938 | ||
| BC171894.1 | - | 3 | 380 | |||
| BC171902.1 | - | 3 | 384 | 9054556 | ||
| BM552707.1 | - | 3 | 431 | |||
| CR541986.1 | - | 3 | 260 | |||
| CR542015.1 | - | 3 | 260 | |||
| CX166352.1 | - | 3 | 445 | |||
| L16991.1 | - | 3 | 302 | |||
| X54729.1 | - | 3 | 284 | |||

DTYMK
Genomic Map