siRNA Details
| Gene Symbol: SP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017371.4 | NP_001017371.3 | 2 | 1092 | ||
| NM_001172712.1 | NP_001166183.1 | 4 | 1434 | |||
| NM_003111.4 | NP_003102.1 | 4 | 1443 | |||
| GenBank mRNA | AB209334.1 | - | 1 | 556 | ||
| AK304199.1 | - | 4 | 1015 | |||
| AY070137.1 | - | 2 | 1092 | |||
| AY441957.1 | - | 4 | 1443 | |||
| AY441958.1 | - | 2 | 782 | |||
| BC126414.1 | - | 4 | 965 | 8992130 | ||
| BC143928.1 | - | 4 | 956 | 9052446 | ||
| M97191.2 | - | 2 | 703 | |||
| X68560.1 | - | 2 | 661 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001098425, NM_001018042, AK088238, BC079874, BC027797, and others.

SP3
Genomic Map