siRNA Details
| Gene Symbol: TULP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289395.1 | NP_001276324.1 | 5 | 419 | ||
| NM_003322.4 | NP_003313.3 | 6 | 578 | |||
| GenBank mRNA | AB593123.1 | - | 6 | 578 | ||
| AB593124.1 | - | 6 | 564 | |||
| BC032714.1 | - | 5 | 404 | 5495136 | ||
| BC065261.1 | - | 5 | 387 | 6144003 | ||
| CN355966.1 | - | 2 | 387 | |||
| U82468.1 | - | 6 | 570 | |||

TULP1
Genomic Map