siRNA Details
Gene Symbol: TULP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003324.4 | NP_003315.2 | 11 | 2427 | ||
GenBank mRNA | BC032587.1 | - | 12 | 2231 | 5501868 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TULP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003324.4 | NP_003315.2 | 11 | 2427 | ||
GenBank mRNA | BC032587.1 | - | 12 | 2231 | 5501868 |