siRNA Details
| Gene Symbol: ZNF35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003420.3 | NP_003411.3 | 3 | 498 | ||
| XM_017007129.1 | XP_016862618.1 | 3 | 472 | |||
| XM_017007130.1 | XP_016862619.1 | 3 | 493 | |||
| XM_017007131.1 | XP_016862620.1 | 3 | 494 | |||
| GenBank mRNA | AK314821.1 | - | 3 | 497 | ||
| BC013597.1 | - | 3 | 476 | 4186299 | ||
| BP237260.1 | - | 3 | 498 | |||
| BT006962.1 | - | 2 | 253 | |||
| X07289.1 | - | 3 | 466 | |||

ZNF35
Genomic Map