siRNA Details
| Gene Symbol: ZNF132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003433.3 | NP_003424.3 | 3 | 714 | ||
| GenBank mRNA | AK057460.1 | - | 3 | 714 | ||
| AK313895.1 | - | 1 | 196 | |||
| BC109107.1 | - | 3 | 508 | 40035328 | ||
| BC109108.1 | - | 3 | 508 | 40035329 | ||
| U09411.1 | - | 1 | 460 | |||

ZNF132
Genomic Map