siRNA Details
| Gene Symbol: MAD2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC000356.2 | - | 5 | 759 | 2964388 | |
| BC005945.1 | - | 5 | 752 | 4132892 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAD2L1| Gene Symbol: MAD2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC000356.2 | - | 5 | 759 | 2964388 | |
| BC005945.1 | - | 5 | 752 | 4132892 | ||