siRNA Details
| Gene Symbol: KLF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB015132.1 | - | 4 | 1347 | ||
| HF546205.1 | - | 4 | 655 | |||
| HF546206.1 | - | 3 | 594 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLF7| Gene Symbol: KLF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB015132.1 | - | 4 | 1347 | ||
| HF546205.1 | - | 4 | 655 | |||
| HF546206.1 | - | 3 | 594 | |||