siRNA Details
| Gene Symbol: MAGEB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002367.3 | NP_002358.1 | 1 | 1253 | ||
| GenBank mRNA | BC032852.2 | - | 1 | 1141 | 5267183 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAGEB4| Gene Symbol: MAGEB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002367.3 | NP_002358.1 | 1 | 1253 | ||
| GenBank mRNA | BC032852.2 | - | 1 | 1141 | 5267183 | |