siRNA Details
| Gene Symbol: MAGOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002370.3 | NP_002361.1 | 5 | 670 | ||
| GenBank mRNA | AF035940.1 | - | 5 | 573 | ||
| AF067173.1 | - | 5 | 596 | |||
| AF086246.1 | - | 3 | 309 | |||
| AK297895.1 | - | 4 | 476 | |||
| BC018211.1 | - | 5 | 582 | 3861094 | ||

MAGOH
Genomic Map