siRNA Details
| Gene Symbol: ZNF213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134655.1 | NP_001128127.1 | 6 | 2778 | ||
| NM_004220.2 | NP_004211.1 | 6 | 2797 | |||
| NR_104432.1 | 6 | 2861 | ||||
| GenBank mRNA | AF017433.1 | - | 7 | 2715 | ||
| AK055337.1 | - | 6 | 2758 | |||
| BC007287.1 | - | 6 | 2715 | 3347511 | ||
| BC080572.1 | - | 1 | 492 | |||

ZNF213
Genomic Map