siRNA Details
| Gene Symbol: TRIP13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166260.1 | NP_001159732.1 | 4 | 566 | ||
| NM_004237.3 | NP_004228.1 | 4 | 566 | |||
| XM_011514163.2 | XP_011512465.1 | 4 | 536 | |||
| GenBank mRNA | AK310376.1 | - | 4 | 536 | ||
| BC000404.2 | - | 4 | 531 | 2822981 | ||
| BC019294.1 | - | 4 | 531 | 2822981 | ||
| BG722761.1 | - | 4 | 568 | |||
| CR456744.1 | - | 4 | 421 | |||
| L40384.1 | - | 4 | 427 | |||
| U96131.1 | - | 4 | 466 | |||

TRIP13
Genomic Map