siRNA Details
| Gene Symbol: FOXA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004497.2 | NP_004488.2 | 2 | 330 | ||
| GenBank mRNA | AK223473.1 | - | 2 | 259 | ||
| BC016024.1 | - | 2 | 330 | 3908052 | ||
| BT006720.1 | - | 2 | 133 | |||
| L12141.1 | - | 2 | 133 | |||

FOXA3
Genomic Map