siRNA Details
| Gene Symbol: FOXK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004514.3 | NP_004505.2 | 9 | 2777 | ||
| GenBank mRNA | AK096800.1 | - | 4 | 1917 | ||
| AK098140.1 | - | 1 | 1595 | |||
| U58196.1 | - | 9 | 2788 | |||
| U58197.1 | - | 10 | 3212 | |||
| U58198.1 | - | 5 | 1910 | |||

FOXK2
Genomic Map