siRNA Details
| Gene Symbol: MCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085377.1 | NP_001078846.1 | 19 | 3955 | ||
| NM_002387.2 | NP_002378.1 | 17 | 3499 | |||
| XM_005271991.2 | XP_005272048.1 | 18 | 3599 | |||
| XM_017009473.1 | XP_016864962.1 | 20 | 3620 | |||
| XM_017009474.1 | XP_016864963.1 | 18 | 6285 | |||
| GenBank mRNA | BX537952.1 | - | 18 | 2970 | ||
| BX647710.1 | - | 20 | 3891 | |||
| M62397.1 | - | 18 | 3189 | |||

MCC
Genomic Map