siRNA Details
| Gene Symbol: MAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031804.2 | NP_001026974.1 | 1 | 6241 | ||
| NM_005360.4 | NP_005351.2 | 2 | 2010 | |||
| GenBank mRNA | AF055377.1 | - | 2 | 1994 | ||
| AF447709.1 | - | 1 | 66 | |||
| BC081542.1 | - | 1 | 5534 | 30347784 | ||

MAF
Genomic Map