siRNA Details
| Gene Symbol: PHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005392.3 | NP_005383.3 | 4 | 583 | ||
| XM_005252051.2 | XP_005252108.1 | 4 | 577 | |||
| XM_006717143.2 | XP_006717206.1 | 4 | 577 | |||
| GenBank mRNA | AB014562.2 | - | 3 | 476 | ||
| AL834263.1 | - | 4 | 434 | |||
| BC152437.1 | - | 3 | 476 | |||
| BU507256.1 | - | 468 | ||||
| CB990295.1 | - | 4 | 611 | |||

PHF2
Genomic Map