siRNA Details
Gene Symbol: TCF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005650.3 | NP_005641.1 | 2 | 516 | ||
NM_181492.2 | NP_852469.1 | 1 | 415 | |||
XM_005261722.3 | XP_005261779.1 | 2 | 1719 | |||
XM_006724313.3 | XP_006724376.1 | 2 | 700 | |||
XM_011530353.2 | XP_011528655.1 | 2 | 700 | |||
XM_011530354.2 | XP_011528656.1 | 2 | 700 | |||
GenBank mRNA | KF851355.1 | - | 2 | 516 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013836, NM_001114140, BC171946, AY007594, BC138038.
Panther Classification:
Gene Ontology Categories:
