siRNA Details
| Gene Symbol: TRIM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199573.1 | NP_001186502.1 | 2 | 762 | ||
| NM_006074.4 | NP_006065.2 | 2 | 762 | |||
| GenBank mRNA | AK290434.1 | - | 2 | 492 | ||
| AK298715.1 | - | 3 | 442 | |||
| AK308796.1 | - | 2 | 478 | |||
| BC022281.1 | - | 2 | 569 | 4718024 | ||
| BC035582.1 | - | 2 | 535 | 5583800 | ||
| CD621768.1 | - | 2 | 666 | |||
| HQ842635.1 | - | 1 | 383 | |||
| X82200.1 | - | 2 | 505 | |||

TRIM22
Genomic Map