siRNA Details
Gene Symbol: PA2G4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006191.2 | NP_006182.2 | 13 | 1988 | ||
GenBank mRNA | U59435.1 | - | 13 | 1667 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PA2G4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006191.2 | NP_006182.2 | 13 | 1988 | ||
GenBank mRNA | U59435.1 | - | 13 | 1667 |