siRNA Details
| Gene Symbol: MMP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002428.3 | NP_002419.1 | 10 | 3011 | ||
| GenBank mRNA | BC006412.1 | - | 10 | 2131 | ||
| BC036495.1 | - | 10 | 3017 | 5263145 | ||
| BC055428.1 | - | 10 | 3012 | 4811572 | ||
| D85510.1 | - | 7 | 1741 | |||
| Z48482.1 | - | 10 | 2274 | |||

MMP15
Genomic Map