siRNA Details
| Gene Symbol: ZNF75D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007131.4 | NP_009062.2 | 4, 5 | 3306 | ||
| XM_005262469.1 | XP_005262526.1 | 4, 5 | 1835 | |||
| XM_011531391.1 | XP_011529693.1 | 5, 6 | 2022 | |||
| XM_011531396.2 | XP_011529698.1 | 3, 4 | 2799 | |||
| GenBank mRNA | AK092347.1 | - | 1, 2 | 195 | ||
| AK290215.1 | - | 3, 4 | 1010 | |||
| AL831984.2 | - | 4, 5 | 3251 | |||
| AL832801.1 | - | 3, 4 | 793 | |||
| BC109100.2 | - | 3, 4 | 825 | 40034784 | ||

ZNF75D
Genomic Map