siRNA Details
| Gene Symbol: ZNF79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286696.1 | NP_001273625.1 | 3, 4 | 747 | ||
| NM_001286697.1 | NP_001273626.1 | 4, 5 | 749 | |||
| NM_001322260.1 | NP_001309189.1 | 4, 5 | 878 | |||
| NM_007135.2 | NP_009066.2 | 3, 4 | 640 | |||
| XM_006717279.3 | XP_006717342.1 | 4, 5 | 848 | |||
| GenBank mRNA | AK054606.1 | - | 3, 4 | 632 | ||
| AK302284.1 | - | 3, 4 | 689 | |||
| AK316233.1 | - | 4, 5 | 685 | |||
| BC062309.1 | - | 3, 4 | 595 | 3944657 | ||

ZNF79
Genomic Map