siRNA Details
| Gene Symbol: ZNF80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007136.3 | NP_009067.2 | 1 | 1018 | ||
| XM_017007132.1 | XP_016862621.1 | 1 | 706 | |||
| XM_017007133.1 | XP_016862622.1 | 1 | 706 | |||
| XM_017007134.1 | XP_016862623.1 | 1 | 706 | |||
| XM_017007135.1 | XP_016862624.1 | 1 | 706 | |||
| XM_017007136.1 | XP_016862625.1 | 1 | 706 | |||
| GenBank mRNA | BC069606.1 | - | 1 | 712 | 7262407 | |
| BC069826.1 | - | 1 | 711 | 7262419 | ||
| BC129925.1 | - | 1 | 541 | 40121991 | ||
| X65233.1 | - | 1 | 1026 | |||

ZNF80
Genomic Map