siRNA Details
| Gene Symbol: ZNF146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099638.1 | NP_001093108.1 | 3 | 1864 | ||
| NM_001099639.1 | NP_001093109.1 | 3 | 1823 | |||
| NM_007145.2 | NP_009076.2 | 4 | 1938 | |||
| XM_005259214.2 | XP_005259271.1 | 4 | 1763 | |||
| XM_017027244.1 | XP_016882733.1 | 3 | 1389 | |||
| XM_017027245.1 | XP_016882734.1 | 4 | 1394 | |||
| XM_017027246.1 | XP_016882735.1 | 3 | 1293 | |||
| XM_017027247.1 | XP_016882736.1 | 3 | 1355 | |||
| GenBank mRNA | AK311486.1 | - | 1 | 408 | ||
| AK313460.1 | - | 1 | 876 | |||
| BC065568.1 | - | 4 | 1407 | |||
| BC110500.1 | - | 1 | 1164 | 40028224 | ||
| BC110501.1 | - | 1 | 1164 | 40028226 | ||
| X70394.1 | - | 2 | 1271 | |||

ZNF146
Genomic Map