siRNA Details
| Gene Symbol: TARDBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007375.3 | NP_031401.1 | 3 | 437 | ||
| XM_017000863.1 | XP_016856352.1 | 3 | 425 | |||
| XM_017000864.1 | XP_016856353.1 | 3 | 425 | |||
| XM_017000865.1 | XP_016856354.1 | 3 | 425 | |||
| XM_017000866.1 | XP_016856355.1 | 3 | 425 | |||
| XM_017000867.1 | XP_016856356.1 | 3 | 425 | |||
| XM_017000868.1 | XP_016856357.1 | 3 | 425 | |||
| GenBank mRNA | AK222754.1 | - | 3 | 375 | ||
| AK299454.1 | - | 3 | 188 | |||
| AK312416.1 | - | 3 | 405 | |||
| AL050265.1 | - | 3 | 391 | |||
| BC001487.2 | - | 3 | 390 | |||
| BC071657.1 | - | 3 | 388 | 5498250 | ||
| BC095435.1 | - | 3 | 389 | 30389805 | ||
| BI464234.1 | - | 4 | 351 | |||
| CD703666.1 | - | 3 | 463 | |||
| CR533534.1 | - | 2 | 303 | |||
| EF434181.1 | - | 2 | 303 | |||
| EF434182.1 | - | 2 | 303 | |||
| HQ628634.1 | - | 3 | 212 | |||
| HQ628635.1 | - | 2 | 303 | |||
| HQ628636.1 | - | 2 | 303 | |||
| HQ628637.1 | - | 2 | 303 | |||
| U23731.1 | - | 2 | 303 | |||

TARDBP
Genomic Map