siRNA Details
| Gene Symbol: PTRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012232.5 | NP_036364.2 | 2 | 737 | ||
| GenBank mRNA | AF312393.1 | - | 2 | 934 | ||
| AK000715.1 | - | 2 | 223 | |||
| AK092406.1 | - | 3 | 538 | |||
| AK298072.1 | - | 3 | 618 | |||
| AK298563.1 | - | 3 | 592 | |||
| AK314389.1 | - | 2 | 673 | |||
| BC066123.1 | - | 2 | 673 | 5269172 | ||
| BC073759.1 | - | 264 | 6572478 | |||

PTRF
Genomic Map