siRNA Details
| Gene Symbol: PTRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012232.5 | NP_036364.2 | 2 | 1835 | ||
| GenBank mRNA | AF312393.1 | - | 2 | 2032 | ||
| AK000715.1 | - | 2 | 1321 | |||
| AK074512.1 | - | 1036 | ||||
| AK092406.1 | - | 4 | 1474 | |||
| BC004295.1 | - | 1 | 1045 | |||
| BC008849.1 | - | 2 | 1118 | 4140111 | ||
| BC066123.1 | - | 2 | 1771 | 5269172 | ||
| BC073759.1 | - | 2 | 1657 | 6572478 | ||

PTRF
Genomic Map