siRNA Details
| Gene Symbol: ZMYM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039650.2 | NP_001034739.1 | 5 | 948 | ||
| NM_001142684.1 | NP_001136156.1 | 5 | 948 | |||
| XM_005266593.3 | XP_005266650.1 | 5 | 1335 | |||
| XM_005266594.2 | XP_005266651.1 | 4 | 763 | |||
| XM_006719894.2 | XP_006719957.1 | 5 | 1058 | |||
| XM_011535309.1 | XP_011533611.1 | 5 | 1307 | |||
| GenBank mRNA | AF161535.1 | - | 2 | 539 | ||
| AJ133352.1 | - | 5 | 836 | |||
| AJ133355.1 | - | 4 | 1076 | |||
| BU853478.1 | - | 1 | 44 | |||

ZMYM5
Genomic Map