siRNA Details
| Gene Symbol: TRIM35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304495.1 | NP_001291424.1 | 4 | 1287 | ||
| NM_171982.4 | NP_741983.2 | 6 | 1406 | |||
| XM_005273452.3 | XP_005273509.1 | 5 | 1378 | |||
| XM_011544453.1 | XP_011542755.1 | 7 | 1434 | |||
| GenBank mRNA | AB029021.1 | - | 6 | 1356 | ||
| AF492463.1 | - | 6 | 1404 | |||
| BC018337.1 | - | 6 | 1516 | |||
| BC069226.1 | - | 6 | 1348 | 6428667 | ||

TRIM35
Genomic Map