siRNA Details
| Gene Symbol: HIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015094.2 | NP_055909.2 | 3 | 4485 | ||
| XM_011530007.2 | XP_011528309.1 | 3 | 6759 | |||
| XM_011530008.2 | XP_011528310.1 | 3 | 4675 | |||
| XM_011530009.2 | XP_011528311.1 | 4 | 6121 | |||
| XM_011530010.2 | XP_011528312.1 | 3 | 4671 | |||
| XM_017028669.1 | XP_016884158.1 | 2 | 4388 | |||
| GenBank mRNA | AB028943.2 | - | 3 | 4485 | ||
| AK023021.1 | - | 1 | 2286 | |||
| BC094787.1 | - | 3 | 4418 | 30531041 | ||

HIC2
Genomic Map