siRNA Details
| Gene Symbol: ZNF777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015694.2 | NP_056509.2 | 5 | 1460 | ||
| XM_005249980.3 | XP_005250037.1 | 5 | 1446 | |||
| XM_011516055.2 | XP_011514357.1 | 5 | 1470 | |||
| GenBank mRNA | AB033111.1 | - | 5 | 1374 | ||
| AK074515.1 | - | 3 | 346 | |||
| AK075032.1 | - | 4 | 557 | |||
| BC023985.2 | - | 4 | 524 | 2820826 | ||
| BC105968.1 | - | 4 | 794 | |||

ZNF777
Genomic Map