siRNA Details
| Gene Symbol: ZNF691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242739.1 | NP_001229668.1 | 4 | 1305 | ||
| NM_015911.3 | NP_056995.1 | 2 | 1004 | |||
| XM_006710671.3 | XP_006710734.1 | 3 | 1121 | |||
| XM_017001400.1 | XP_016856889.1 | 3 | 1450 | |||
| XM_017001401.1 | XP_016856890.1 | 2 | 3987 | |||
| XM_017001402.1 | XP_016856891.1 | 3 | 2451 | |||
| XM_017001403.1 | XP_016856892.1 | 2 | 4987 | |||
| GenBank mRNA | AF131739.1 | - | 2 | 968 | ||
| AK296201.1 | - | 3 | 1393 | |||
| BC000157.2 | - | 2 | 1047 | 2901198 | ||

ZNF691
Genomic Map