siRNA Details
| Gene Symbol: ZNF581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016535.3 | NP_057619.1 | 2 | 335 | ||
| XM_006723240.3 | XP_006723303.1 | 2 | 358 | |||
| XM_017026867.1 | XP_016882356.1 | 2 | 268 | |||
| GenBank mRNA | AF151023.2 | - | 2 | 335 | ||
| AK026203.1 | - | 3 | 272 | |||
| AK315603.1 | - | 2 | 306 | |||
| BC001219.1 | - | 2 | 304 | 3528677 | ||
| BC010097.2 | - | 2 | 307 | 3504825 | ||
| BC071620.1 | - | 2 | 434 | 4446945 | ||
| BC071845.1 | - | 2 | 289 | 4764567 | ||

ZNF581
Genomic Map