siRNA Details
| Gene Symbol: NDUFS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002496.3 | NP_002487.1 | 7 | 662 | ||
| XM_005274013.1 | XP_005274070.1 | 7 | 759 | |||
| XM_005274014.2 | XP_005274071.1 | 7 | 786 | |||
| XM_011545053.2 | XP_011543355.1 | 7 | 697 | |||
| GenBank mRNA | AK223114.1 | - | 7 | 652 | ||
| AK314546.1 | - | 7 | 587 | |||
| BC119753.1 | - | 3 | 219 | |||
| BC119754.2 | - | 7 | 591 | 40119290 | ||
| U65579.1 | - | 7 | 585 | |||

NDUFS8
Genomic Map