siRNA Details
| Gene Symbol: KDM3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146688.1 | NP_001140160.1 | 3, 4 | 410 | ||
| NM_018433.5 | NP_060903.2 | 3, 4 | 679 | |||
| XM_006712051.3 | XP_006712114.1 | 2, 3 | 382 | |||
| XM_017004493.1 | XP_016859982.1 | 3, 4 | 698 | |||
| XM_017004494.1 | XP_016859983.1 | 3, 4 | 468 | |||
| GenBank mRNA | AB018285.2 | - | 3, 4 | 442 | ||
| AL832150.1 | - | 3, 4 | 445 | |||
| BC038297.2 | - | 3, 4 | 456 | |||
| BG723267.1 | - | 3, 4 | 461 | |||
| BX640698.1 | - | 3, 4 | 480 | |||
| BX647271.1 | - | 3, 4 | 448 | |||
| BX647752.1 | - | 3, 4 | 508 | |||
| BX648841.1 | - | 3, 4 | 411 | |||
| BX649109.1 | - | 3, 4 | 449 | |||
| CR749581.1 | - | 3, 4 | 447 | |||

KDM3A
Genomic Map