siRNA Details
Gene Symbol: LMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001395.2 | NP_001001395.1 | 2 | 453 | ||
NM_001243609.1 | NP_001230538.1 | 3 | 520 | |||
NM_001243610.1 | NP_001230539.1 | 3 | 292 | |||
NM_001243611.1 | NP_001230540.1 | 2 | 689 | |||
NM_001243612.1 | NP_001230541.1 | 2 | 377 | |||
NM_018640.4 | NP_061110.2 | 2 | 711 | |||
NR_045012.1 | 3 | 520 | ||||
XM_006719110.3 | XP_006719173.1 | 3 | 528 | |||
XM_006719111.2 | XP_006719174.1 | 2 | 571 | |||
XM_011520762.1 | XP_011519064.1 | 3 | 419 | |||
XM_011520763.1 | XP_011519065.1 | 3 | 326 | |||
XM_017019643.1 | XP_016875132.1 | 2 | 674 | |||
GenBank mRNA | AB044745.1 | - | 2 | 457 | ||
AB044746.1 | - | 4 | 358 | |||
AK095595.1 | - | 2 | 329 | |||
AK294474.1 | - | 2 | 377 | |||
AK294651.1 | - | 2 | 366 | |||
AK308594.1 | - | 3 | 431 | |||
AK308826.1 | - | 4 | 396 | |||
AK308896.1 | - | 3 | 332 | |||
AK312418.1 | - | 2 | 595 | |||
AK316021.1 | - | 2 | 313 | |||
BC026311.1 | - | 2 | 607 | 4799836 | ||
BC050085.1 | - | 3 | 293 | 5274811 | ||
BI553855.1 | - | 2 | 350 | |||
DA780743.1 | - | 2 | 454 |
